Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500511

Sigma-Aldrich

Anti-CKI-ε antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CK1-ε, CKI-ε, CKIe, CSNK1E, Casein kinase I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 47 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSNK1E(1454)

Related Categories

General description

Casein kinase 1 ε (CSNK1E) is part of the serine/threonine protein kinase family. The gene encoding it is localized on human chromosome 22q13.1. Anti-CKI-ε Antibody detects endogenous levels of total CKI-ε protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CKI-epsilon.

Immunogen Range: 276-325

Biochem/physiol Actions

Casein kinase 1 ε (CSNK1E) phosphorylates proteins of the Wnt signaling cascade. It modulates tumor growth and cell division in salivary gland cancer, pancreatic and colon adenocarcinoma cells. CSNK1E phosphorylates dopamine- and cAMP-regulated neuronal phosphoprotein (DARPP-32) and modulates dopamine receptor signaling. It also interacts with proteins involved in circadian rhythm.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of genes associated with tumorigenesis of meibomian cell carcinoma by microarray analysis.
Kumar A, et. al
Genomics, 90(5), 559-566 (2007)
Arun Kumar et al.
Genomics, 90(5), 559-566 (2007-09-25)
Meibomian cell carcinoma (MCC) is a malignant tumor of the meibomian glands located in the eyelids. No information exists on the cytogenetic and genetic aspects of MCC. There is no report on the gene expression profile of MCC. Thus there
Dopaminergic pathway polymorphisms and heroin addiction: further support for association of CSNK1E variants.
Levran O
Pharmacogenomics, 15(16), 2001-2009 (2014)
Association between a casein kinase 1 e gene polymorphism and schizophrenia in a Chinese Han population.
Huang Y
Journal of Molecular Neuroscience, 47(3), 470-474 (2012)
Jose Luis Lopez-Guerra et al.
Oncotarget, 6(30), 30343-30356 (2015-09-04)
Reliable biological markers that predict breast cancer (BC) outcomes after multidisciplinary therapy have not been fully elucidated. We investigated the association between casein kinase 1 epsilon (CK1ε) and the risk of recurrence in patients with BC. Using 168 available tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service