Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500206

Sigma-Aldrich

Anti-ADCY5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ATP pyrophosphate-lyase, CYA6, Ca(2 )-inhibitable adenylyl cyclase, KIAA0422, adenylate cyclase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€391.00

€391.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€391.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€391.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 130 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADCY5(112)

General description

Anti-ADCY5 Antibody detects endogenous levels of total ADCY5 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ADCY5/6.

Immunogen Range: 931-980

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Hayakawa et al.
The journal of physiological sciences : JPS, 72(1), 2-2 (2022-02-13)
We recently reported a positive relationship between occlusal disharmony and cardiovascular disease via activation of β-adrenergic signaling in mice. Furthermore, inhibition of type 5 adenylyl cyclase (AC5), a major cardiac subtype in adults, protects the heart against oxidative stress. Here
Michinori Tsunoda et al.
The journal of physiological sciences : JPS, 73(1), 18-18 (2023-08-10)
In this work, we examined the involvement of type 5 adenylyl cyclase (AC5) in cardiac dysfunction induced in mice given Porphyromonas gingivalis lipopolysaccharide (PG-LPS) at a dose equivalent to the circulating levels in periodontitis (PD) patients. Cardiac function was significantly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service