Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300471

Sigma-Aldrich

Anti-CTNNB1 (Ab-37) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CTNNB antibody produced in rabbit, Anti-DKFZp686D02253 antibody produced in rabbit, Anti-FLJ25606 antibody produced in rabbit, Anti-FLJ37923 antibody produced in rabbit, Anti-catenin (cadherin-associated protein), beta 1, 88kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€400.00

€400.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€400.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€400.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~92 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(I-H-S-G-A)

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)

Immunogen

Peptide sequence around aa. 35-39 (I-H-S-G-A), according to the protein CTNNB1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Involved in the regulation of cell adhesion and in signal transduction through the Wnt pathway.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service