Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300396

Sigma-Aldrich

Anti-CHUK (Ab-23) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IKBKA antibody produced in rabbit, Anti-IKK-alpha antibody produced in rabbit, Anti-IKK1 antibody produced in rabbit, Anti-IKKA antibody produced in rabbit, Anti-conserved helix-loop-helix ubiquitous kinase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€340.00

€340.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€340.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€340.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~85 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(L-G-T-G-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHUK(1147)

Related Categories

Immunogen

Peptide sequence around aa. 21-25 (L-G-T-G-G), according to the protein CHUK.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Acts as part of the IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. As part of the non-canonical pathway of NF-kappa-B activation, the MAP3K14-activated CHUK/IKKA homodimer phosphorylates NFKB2/p100 associated with RelB, inducing its proteolytic processing to NFKB2/p52 and the formation of NF-kappa-B RelB-p52 complexes. Also phosphorylates NCOA3. Phosphorylates 'Ser-10' of histone H3 at NF-kappa-B-regulated promoters during inflammatory responses triggered by cytokines.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service