Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300270

Sigma-Aldrich

Anti-phospho-AURKA (pThr288) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AIK antibody produced in rabbit, Anti-ARK1 antibody produced in rabbit, Anti-AURA antibody produced in rabbit, Anti-AURORA2 antibody produced in rabbit, Anti-aurora kinase A antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€490.00

€490.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€490.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€490.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~48 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100

isotype

IgG

immunogen sequence

(R-T-TP-L-M)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr288)

Gene Information

human ... AURKA(6790)

Related Categories

Immunogen

Peptide sequence around phosphorylation site of threonine 288 (R-T-T(p)-L-M), according to the protein AURKA.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Contributes to the regulation of cell cycle progression. Required for normal mitosis. Associates with the centrosome and the spindle microtubules during mitosis and functions in centrosome maturation, spindle assembly, maintenance of spindle bipolarity, centrosome separation and mitotic checkpoint control. Phosphorylates numerous target proteins, including ARHGEF2, BRCA1, KIF2A, NDEL1, PARD3, PLK1 and BORA. Regulates KIF2A tubulin depolymerase activity By similarity. Required for normal axon formation. Plays a role in microtubule remodeling during neurite extension. Important for microtubule formation and/or stabilization.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Yun Yang et al.
Head & neck, 41(7), 2239-2248 (2019-02-02)
Chemotherapy resistance was an important tumor metastasis mechanism. Cell Counting Kit-8 assay and plate colony formation assay were applied to examine the proliferation of laryngeal squamous cell carcinoma (LSCC). Immunofluorescent staining and Western blotting were carried out to show the
Suqin Shen et al.
Cell cycle (Georgetown, Tex.), 18(18), 2228-2238 (2019-07-31)
Aurora-A is a serine/threonine kinase, which is overexpressed in multiple human cancers and plays a key role in tumorigenesis and tumor development. In this study, we found that the receptor of activated C-kinase1 (RACK1), an important regulator of biological functions
Seul Kim et al.
Toxicology in vitro : an international journal published in association with BIBRA, 59, 115-125 (2019-04-14)
Bisphenol A [BPA, 2,2-bis-(4-hydroxyphenyl)propane] is one of the most prevalent synthetic environmental estrogens; as an endocrine disruptor, it is associated with endocrine-related cancers including breast, ovarian, and prostate. However, the mechanisms by which BPA contributes to carcinogenesis are unclear. This
Se-Chun Liao et al.
Nutrients, 11(6) (2019-06-27)
Curcumin is a polyphenol derived from curcumin longa that exhibits anticancer and anti-inflammatory properties. The consumption of foods at supernutritional levels to obtain health benefits may paradoxically result in negative health outcomes. In the present study, multiple targeting characteristics of
Pratim Chowdhury et al.
Journal of cell science, 131(24) (2018-12-07)
Loss of the gene von Hippel-Lindau (VHL) is associated with loss of primary cilia and is causally linked to elevated levels of Aurora kinase A (AURKA). We developed an image-based high-throughput screening (HTS) assay using a dual-labeling image analysis strategy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service