Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300198

Sigma-Aldrich

Anti-phospho-MAPK9 (pThr183) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JNK-55 antibody produced in rabbit, Anti-JNK2 antibody produced in rabbit, Anti-JNK2A antibody produced in rabbit, Anti-JNK2ALPHA antibody produced in rabbit, Anti-mitogen-activated protein kinase 9 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~46 kDa
~54 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(M-M-TP-P-Y)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr183)

Gene Information

human ... MAPK9(5601)

Related Categories

Immunogen

Peptide sequence around phosphorylation site of threonine 183 (M-M-T(p)-P-Y), according to the protein MAPK9.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Responds to activation by environmental stress and pro-inflammatory cytokines by phosphorylating a number of transcription factors, primarily components of AP-1 such as c-Jun and ATF2 and thus regulates AP-1 transcriptional activity. In T-cells, JNK1 and JNK2 are required for polarized differentiation of T-helper cells into Th1 cells.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wang et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 276(3), 767-774 (2019-01-03)
Recent studies have shown that laryngopharyngeal reflux is associated with chronic rhinosinusitis. Pepsin may be a key factor involved in the injury of nasal mucosal epithelial cells, but the pathogenesis remains unclear. We are to investigate whether a mitogen-activated protein
Nuria Suelves et al.
Molecular neurobiology, 56(2), 935-953 (2018-05-29)
Deficits in striatal brain-derived neurotrophic factor (BDNF) delivery and/or BDNF/tropomyosin receptor kinase B (TrkB) signaling may contribute to neurotrophic support reduction and selective early degeneration of striatal medium spiny neurons in Huntington's disease (HD). Furthermore, we and others have demonstrated
Mengchu Cui et al.
Cell transplantation, 28(5), 568-584 (2019-03-06)
Repetitive transcranial magnetic stimulation (rTMS) is a physical treatment applied during recovery after intracerebral hemorrhage (ICH). With in vivo and in vitro assays, the present study sought to investigate how rTMS influences neural stem cells (NSCs) after ICH and the
Chaobin Lin et al.
Eye (London, England), 33(5), 777-784 (2018-12-19)
Glaucoma is the leading cause of vision loss and blindness in the world. Elucidating the pathogenesis of glaucoma and developing effective treatment should be the priority. Inflammation and oxidative stress play essential roles in glaucoma pathogeneisis. Kaempferol is a natural
Yong-Guo Zhang et al.
Inflammatory bowel diseases, 25(1), 97-110 (2018-10-06)
Vitamin D3 and vitamin D receptor (VDR) are involved in the pathogenesis of inflammatory bowel disease (IBD) and bacterial infection. Claudin-2 is a junction protein that mediates paracellular water transport in epithelia. Elevation of Claudin-2 is associated with active IBD.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service