Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200839

Sigma-Aldrich

Anti-Acetylcholinesterase (AChE) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

AChE

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€126.00
100 μL
€330.00

€126.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€126.00
100 μL
€330.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€126.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1:1000-1:2000 using whole extracts of Jurkat cells.
immunofluorescence: 1:1000-1:20000 using Human A431

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACHE(43)

General description

Acetylcholinesterase (AChE) is a member of the a/ß-hydrolase-fold proteins superfamily which is composed of structurally related proteins with a great diversity in their catalytic, recognition, adhesion and chaperone functions. 3D structure studies of a/ß-hydrolase reviled a variety of folding patterns suggesting associations through oligomerization dependent functions.1 AChE is a tissue specific serine hydrolase expressed in several variants at neuromuscular junctions (NMJs)2, cholinergic brain synapses2-3 or erythrocyte (Red Blood Cells- RBC)4.

Specificity

Anti-Acetylcholinesterase (AChE) antibody specifically recognizes human AChE.

Immunogen

human Acetylcholinesterase, recombinant, expressed in HEK 293 cells

Application

The antibody may be used in various immunochemical techniques including Immunoblotting Immunofluorescence and ELISA. Detection of the AChE band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

In the synaptic cleft, AChE main function is to ensure rapid hydrolysis of acetylcholine (ACh) released from motor nerve ending into choline and acetic acid thus preventing postsynaptic muscle ACh receptors (AChR) hyperactivation and termination of the signal transmission.2-3,5AChE and Butyrylcholinesterase (BChE) inhibitory drugs or peptides are being used in several neurodegenerative diseases including Alzheimer′s disease and pathological muscle weakness such as Myasthenia Gravis. These AChE inhibitors increase synaptic ACh levels and hence improve the cholinergic function in the brain.2-3,5-6 Erythrocytes AChE (AChE-H) is a membrane encored AChE splicing variant found in RBC, it belongs to the glycosylphosphatidylinositol (GPI) protein family and carries the Yta blood group antigen. Erythrocytes AChE can be found in plasma circulating vesicles originated from aged erythrocytes, and therefore is considered as a biomarker for RBC membrane integrity. Furthermore, Erythrocytes AChE activity is also a marker for inflammation and other diseases including essential hypertension, glaucoma, ALS, neurotoxicity, and Hirschsprung′s disease.4,7-8

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service