Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200790

Sigma-Aldrich

Anti-Human IgG (Fc specific)-Peroxidase antibody, Mouse monoclonal

clone GG-7, purified from hybridoma cell culture

Synonym(s):

Anti-Human immunoglobulin G

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€407.00

€407.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
€407.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€407.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

GG-7, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

direct ELISA: 1:40,000-1:80,000 using 2.5 μg/mL human IgG1 for coating

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgGs are the most common Immunoglobulins isotype in blood, lymph fluid, cerebrospinal fluid, and peritoneal fluid and a key players in the humoral immune response. IgGs include four subclasses (IgG1, IgG2, IgG3, and IgG4), they consist of a variable Fab fragment (which includes the antibody recognition site), and a conserved Fc fragment. The IgG subclasses differ in their physical and chemical properties, their distribution pattern is found to be age-dependent and every subclass has a specific biological function. IgG deficiencies are often associated with various diseases.

The Fc fragment has various important functions such as complement fixation, site for rheumatoid factor (autoantibody directed to Fc) attachment, passage through the placental membrane and protein A binding. A certain population of lymphocytes also possess an "Fc receptor". These functions indicate the importance of immunoreagents specific for the Fc fragment of Human IgG.

Monoclonal Anti-Human IgG (Fc specific) is specific for the Fc fragment of Human IgG and is observed with all human IgG subclasses but not with the Fab fragment of human IgG, purified light chains, human IgA or human IgM.

Immunogen

Purified human IgG myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads.

Application

The antibody is recommended to use in various immunological techniques, including ELISA.

Physical form

Supplied as a lyophilized powder

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service