Skip to Content
Merck
All Photos(2)

Documents

SAB4200446

Sigma-Aldrich

Anti-CD3 ζ chain antibody, Mouse monoclonal

clone ZT-10, purified from hybridoma cell culture

Synonym(s):

Anti-CD247, Anti-CD3-eta, Anti-CD3Z, Anti-CD3h, Anti-T3Z, Anti-TCRZ, Monoclonal Anti-CD3 ζ chain antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ZT-10, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 1.5-3.0 μg/test using Jurkat cells
indirect immunofluorescence: 2.5-5.0 μg/mL using Jurkat cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... CD247(12503)

General description

Monoclonal Anti-CD3ζ chain (mouse IgG2b isotype) is derived from the hybridoma ZT-10 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to amino acids 79-87 of mouse CD3 ζ chain conjugated to KLH. CD3ζ, a 16-kDa transmembrane protein, consists of a short extracellular domain and a long intracellular domain including immunoreceptor tyrosine-based activation motif (ITAM). It is a subunit of T cell receptor chains (TCR), which is expressed by T cells and natural killer (NK) cells.
The CD3ζ (CD247) gene is mapped to human chromosome 1q24.2.

Specificity

Monoclonal Anti-CD3ζ chain recognizes human and mouse CD3ζ chain.

Application

Monoclonal Anti-CD3 ζ chain antibody produced in mouse has been used in immunocytochemistry and flow cytometry.

Biochem/physiol Actions

CD3ζ regulates immunity, neuronal development and synaptic plasticity. It is vital for T cell receptor chains (TCR) complex assembly and expression. Downregulation of TCR ζ-chain expression and impairment of T-cell function linked to systemic lupus erythematosus (SLE), rheumatoid arthritis, cancer and infectious diseases. Altered expression and function of CD3ζ in T cells is associated with hematological diseases.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative

Storage and Stability

For extended storage, freeze at −20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koichiro Asano et al.
Allergy, 75(12), 3087-3099 (2020-10-12)
Eosinophilic airway inflammation is one of the cardinal features of allergic airway diseases such as atopic asthma and allergic rhinitis. These childhood-onset conditions are mediated by allergen and allergen-specific IgE and often accompanied by other allergic diseases including food allergy
Alterations in the expression pattern of TCR zeta chain in T cells from patients with hematological diseases
Li Y
Hematology, 13(5), 267-275 (2008)
Cutting edge: CD3 ITAM diversity is required for optimal TCR signaling and thymocyte development
Bettini ML, et al.
Journal of Immunology, 199(5), 1555-1560 (2017)
T cell CD3zeta deficiency enables multiorgan tissue inflammation
Deng GM, et al.
Journal of immunology (Baltimore, Md. : 1950), 191(7), 3563-3567 (2013)
Antoine Louveau et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(47), 18672-18685 (2013-11-22)
The immunoreceptor-associated protein CD3ζ is known for its role in immunity and has also been implicated in neuronal development and synaptic plasticity. However, the mechanism by which CD3ζ regulates synaptic transmission remains unclear. In this study, we showed that mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service