Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200247

Sigma-Aldrich

Anti-GP1BA antibody, Mouse monoclonal

clone PM6/40, purified from hybridoma cell culture

Synonym(s):

MonoclonalAnti-BSS, MonoclonalAnti-CD42B, MonoclonalAnti-CD42balpha, MonoclonalAnti-GP1B, MonoclonalAnti-MGC34595, MonoclonalAnti-glycoprotein Ib (platelet), alpha polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PM6/40, monoclonal

form

buffered aqueous solution

mol wt

antigen ~140 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 30-60 μg/mL using human platelet
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GP1BA(2811)

General description

Monoclonal Anti-GP1BA (mouse IgG1 isotype) is derived from the hybridoma PM6/40 produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. GP1BA, also known as glycoprotein Ib (GP Ib) and CD42b, is a platelet surface membrane glycoprotein. GP Ib is a heterodimer, composed of an α and β chain, that are linked by disulfide bonds. The complete receptor complex includes noncovalent association of the α- and β-subunits with platelet glycoprotein IX and platelet glycoprotein V. The glycoprotein Ib platelet subunit α (GP1BA) gene is mapped to human chromosome 17p13.2.

Specificity

Monoclonal Anti-GP1BA recognizes human GP1BA/CD42b.

Immunogen

human platelet membranes.

Application

Anti-GP1BA antibody, Mouse monoclonal is suitable for:
  • immunoblotting
  • fluorescence-activated cell sorting (FACS)
  • immunohistochemistry

Biochem/physiol Actions

Glycoprotein Ib (GP Ib) functions as a receptor for von Willebrand factor (VWF). It regulates the adhesion of blood platelets to damaged blood vessel walls and the subsequent platelet aggregation. It also transmits signals leading to platelet activation, aggregation and secretion. The binding of the GP Ib-IX-V complex to VWF, facilitates initial platelet adhesion to vascular subendothelium after vascular injury, and also initiates signaling events within the platelet that lead to enhanced platelet activation, thrombosis, and hemostasis. The interaction between Gp Ib and VWF was found to induce apoptotic events in platelets, suggesting a mechanism for platelet clearance or some thrombocytopenic disease. Furthermore, several polymorphisms and mutations have been described in this gene, some of which are the cause of Bernard Soulier syndromes and platelet-type von Willebrand disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Canobbio et al.
Cellular signalling, 16(12), 1329-1344 (2004-09-24)
The glycoprotein Ib-V-IX is one of the major adhesive receptors expressed on the surface of circulating platelets. It is composed of four different polypeptides-GPIbalpha, GPIbbeta, GPIX, and GPV-and represents a multifunctional receptor able to interact with a number of ligands
Shi-Zhong Luo et al.
Journal of molecular biology, 382(2), 448-457 (2008-08-05)
As the receptor on the platelet surface for von Willebrand factor, glycoprotein (GP) Ib-IX complex is critically involved in hemostasis and thrombosis. How the complex is assembled from GP Ibalpha, GP Ib beta and GP IX subunits, all of which
Jose A Guerrero et al.
Blood, 114(27), 5541-5546 (2009-10-08)
Platelet-type von Willebrand disease (PT-VWD) is a bleeding disorder of the platelet glycoprotein Ib-IX/von Willebrand factor (VWF) axis caused by mutations in the glycoprotein Ib-IX receptor that lead to an increased affinity with VWF. In this report, platelets from a
Platelets
Encyclopedia of Immunology, 1973-1976 null
Alan T Nurden et al.
British journal of haematology, 165(2), 165-178 (2013-11-30)
Genetic defects of platelets constitute rare diseases that include bleeding syndromes of autosomal dominant, recessive or X-linked inheritance. They affect platelet production, resulting in a low circulating platelet count and changes in platelet morphology, platelet function, or a combination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service