Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501264

Sigma-Aldrich

Anti-Rift Valley Fever Virus Polymerase antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€367.00

€367.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€367.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (Rift Valley Fever virus)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Rift Valley fever virus ... RVFV_sL_gp1(9538294)

General description

Rift Valley Fever Virus Polymerase Antibody: Rift Valley Fever (RFV) virus is an arthropod-borne virus endemic to Africa that infects humans and animals that is transmitted predominantly by mosquitoes. During human infections, symptoms can range from benign fever to severe encephalitis and fatal hepatitis with hemorrhagic fever. The Bunyaviridae family of viruses to which the RVF virus belongs are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity. The three segments are referred to as the L, M, and S segments. The L and M segments are negative polarity and code fore the L-dependent RNA polymerase and glycoprotein precursor respectively. The S segment is of ambisense polarity and encodes the nucleoprotein and non-structural proteins.

Immunogen

RVF virus Polymerase antibody was raised against a 16 amino acid peptide near the amino terminus of the RVF virus Polymerase.The immunogen is located within amino acids 370 - 420 of Rift Valley Fever Virus Polymerase.

Application

RVF virus Polymerase antibody can detect 10 ng RVF virus Polymerase peptide in ELISA at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service