Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501212

Sigma-Aldrich

Anti-West Nile Virus Envelope antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€367.00

€367.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€367.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

West Nile virus

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

West Nile Virus Envelope Antibody: West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins. While the viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway, the highly glycosylated envelope protein plays a major role for WNV entry into target cells as this entry can be inhibited by using a recombinant domain III from the envelope glycoprotein. The WNV receptor has recently been identified as alpha v beta 3 integrin.

Immunogen

West Nile virus envelope antibody was raised against a synthetic peptide corresponding to 14 amino acids at the C-terminus of the West Nile Virus envelope protein.The immunogen is located within amino acids 740 - 790 of West Nile Virus Envelope.

Application

West Nile virus envelope protein antibody can be used for the detection of the West Nile virus envelope protein in ELISA. It will detect 10 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service