Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500604

Sigma-Aldrich

Anti-KLOTHO antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-KL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€504.00

€504.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€504.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€504.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 111 kDa

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KL(9365)

General description

The gene KL (klotho) encodes a protein that has homology to β-glucosidase enzymes. The gene is mapped to human chromosome 13q12. The gene transcript spans a length of 5.2kb. Alternative splicing leads to the generation of two forms, a transmembrane and a secreted form. The transmembrane protein contains 1014 amino acids.

Immunogen

KLOTHO antibody was raised against a 16 amino acid peptide near the center of human KLOTHO.

Application

Anti-KL antibody produced in rabbit has been used as a primary antibody in immunoblot technique.[1]

Biochem/physiol Actions

The KL (klotho) gene is also referred to as an anti-aging gene as it encodes a senescence-related molecule. It regulates fibroblast growth factor (FGF) 23 signaling by functioning as a cofactor or coreceptor. It is involved in the activation of TRPV5 (transient receptor potential cation channel subfamily V member 5), an ion channel, by functioning as a glucuronidase. Klotho affects several intracellular signaling pathways, such as p53/p21, cAMP, protein kinase C (PKC) and Wnt signaling pathways. Mutations in this gene have been associated with premature aging-like phenotypes and shortened life span. Single nucleotide polymorphisms in this gene have been associated with priapism in sickle cell anaemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500604.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Su et al.
International journal of general medicine, 15, 7885-7893 (2022-10-29)
This study focused on renal arteriosclerosis and aimed to explore the relationship between Klotho and SIRT1 by morphological staining, which will help to provide new ideas for the treatment of renal-aging-related diseases and a theoretical basis for the development of
Chiao-Yin Sun et al.
Genes, 11(9) (2020-09-10)
The Klotho gene functions as an anti-aging gene. A previous klotho-knockout mice study indicated that neither male nor female gametocytes could accomplish the first meiotic division. It suggested that Klotho might regulate cell division. In this study, we determined the
Astragaloside IV Attenuates High-Glucose-Induced Impairment in Diabetic Nephropathy by Increasing Klotho Expression via the NF-I?B/NLRP3 Axis.
He, et al.
Journal of Diabetes Research, 2023, 7423661-7423661 (2023)
Agnieszka Olejnik et al.
Journal of cellular and molecular medicine, 24(11), 6448-6458 (2020-04-23)
Restoration of blood flow to ischaemic heart inflicts ischaemia/reperfusion (I/R) injury, which manifests in metabolic and morphological disorders. Klotho is a protein with antioxidative and antiapoptotic activity, and is involved in the regulation of inflammation and fibrosis. The aim of
Qi-Feng Liu et al.
Iranian journal of kidney diseases, 9(4), 291-297 (2015-07-16)
Expression of Klotho is decreased and endoplasmic reticulum (ER) stress is activated in patients with chronic kidney disease. This study aimed to investigate the effect of Klotho protein on ER stress-related apoptosis and renal fibrosis in rates with unilateral ureteral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service