Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500439

Sigma-Aldrich

Anti-IL-33 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Interleukin-33, Anti-NFHEV, Anti-Nuclear factor from high endothelial venules

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€406.00

€406.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1350


Select a Size

Change View
100 μG
€406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€406.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1350

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL33(90865)

Immunogen

IL-33 antibody was raised against a 19 amino acid peptide from near the amino terminus of human IL-33.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500439.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaju Duan et al.
Cytokine, 119, 159-167 (2019-03-27)
Immune cells infiltrating the psoriatic skin secrete high amounts of pro-inflammatory cytokines IL-17, TNF-α, IL-21 and IL-36 resulting in chronic inflammation. However, the exact cellular and molecular mechanisms have not been fully understood. We report here elevation of IL-33 expression
Zhixia Fan et al.
International journal of clinical and experimental pathology, 13(4), 799-809 (2020-05-02)
Psoriasis is an immune-mediated skin disease with abnormal T cells. Regulatory T cells (Treg) are a kind of cell group with immunosuppressive effects. This study aimed to explore the role of Treg cells in the pathogenesis of psoriasis and its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service