Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500336

Sigma-Aldrich

Anti-BAD antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Bcl-2-like 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€359.00

€359.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1569


Select a Size

Change View
100 μG
€359.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€359.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1569

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAD(572)

Immunogen

Bad antibody was raised against a peptide corresponding to 15 amino acids near the C-terminus of human Bad.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500336.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuichi Sawaguchi et al.
Anticancer research, 37(8), 4051-4057 (2017-07-26)
The serine/threonine Pim kinases are overexpressed in various types of solid carcinomas and hematological malignancies, and contribute to regulating cell-cycle progression and cell survival. The aim of this study was to discover a novel pan-Pim kinases inhibitor with potent anti-proliferative
Yuichi Sawaguchi et al.
Frontiers in pharmacology, 12, 672536-672536 (2021-05-21)
Pim kinases are overexpressed in various types of hematological malignancies and solid carcinomas, and promote cell proliferation and survival. Here in this study, we investigated the preclinical profile of novel pan-Pim kinase inhibitors with imidazopyridazine and thiazolidinedione structure. Imidazopyridazine-thiazolidinediones inhibited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service