Skip to Content
Merck
All Photos(3)

Key Documents

SAB2702140

Sigma-Aldrich

Monoclonal Anti-CENPF antibody produced in mouse

Synonym(s):

CENF, PRO1779, hcp-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€420.00

€420.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€420.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€420.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14C10 1D8
monoclonal

form

liquid

mol wt

350 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CENPF(1063)

Immunogen

Protein fragment expressed in E. coli corresponding to amino acids 1759-2093.

Application

For IHC-Fr: Use at an assay dependent dilution. For IHC-P: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:500. Predicted molecular weight: 350 kDa. Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

Mitosin-14C10 recognizes mitosin, a 350kDa phosphoprotein that is specifically involved in mitotic-phase progression. Mitosin is expressed throughout S, G2, and M phases of the cell cycle but is absent in G0 and G1. It associates with the mitotic apparatus during M-phase, and is redistributed from the nucleus to the centromere, spindle, and midbody during M-phase progression. Its usefulness as a proliferation marker is under active investigation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service