Skip to Content
Merck
All Photos(5)

Key Documents

SAB2701838

Sigma-Aldrich

Anti-tp63 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

DNp63, MGC192897, id:ibd3516, tp63

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€347.00

€347.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€347.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

technique(s)

immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... tp63(260407)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 79 and 280 of tp63 according to NP_694518

Application

Suggested starting dilutions are as follows: IHC: 1:100-1:1000, IHC-P: Assay-dependent dilution, WB: 1:500-1:3000, IHC-Wm: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Baraban et al.
Developmental cell, 58(5), 361-375 (2023-02-26)
Despite their barrier function, epithelia can locally lose their integrity to create physiological openings during morphogenesis. The mechanisms driving the formation of these epithelial breaks are only starting to be investigated. Here, we study the formation of the zebrafish nostril
Endika Haro et al.
PLoS genetics, 10(8), e1004468-e1004468 (2014-08-29)
The formation and maintenance of the apical ectodermal ridge (AER) is critical for the outgrowth and patterning of the vertebrate limb. The induction of the AER is a complex process that relies on integrated interactions among the Fgf, Wnt, and
Ju Liu et al.
Cell death and differentiation, 21(11), 1792-1804 (2014-08-26)
Tumor suppressor p53 has a key role in maintaining genomic stability and preventing tumorigenesis through its regulation of cellular stress responses, including apoptosis, cell cycle arrest and senescence. To ensure its proper levels and functions in cells, p53 is tightly
U M Abdel-Motal et al.
Gene therapy, 21(9), 802-810 (2014-06-27)
Topical microbicides are a leading strategy for prevention of HIV mucosal infection to women; however, numerous pharmacokinetic limitations associated with coitally related dosing strategy have contributed to their limited success. Here we test the hypothesis that adeno-associated virus (AAV) mediated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service