Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501761

Sigma-Aldrich

Anti-ZNF703 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

FLJ14299, ZNF503L, hypothetical protein LOC80139, zinc finger protein 703

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, canine, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF703(80139)

General description

ZNF703 (zinc finger protein 703) belongs to the NET (Noc/Nlz, Elbow, and Tlp-1) protein family, and the corresponding gene is localized to human chromosome 8p12.

Immunogen

Peptide with sequence C-ELDKKDQEPKPSPE, from the internal region of the protein sequence according toNP_079345.1

Biochem/physiol Actions

ZNF703 (zinc finger protein 703) is a frequently modified gene in the pan-cancer group. In zebrafish and Drosophila, it plays an essential role in embryonic development. Amplification of this gene leads to loss of cell-cell adhesion, and thus, an increase in proliferation and migration. In patients with head and neck squamous cell carcinomas (HNSCC), the overexpression of this protein is linked with poor overall survival. It functions as an oncogene in luminal B breast cancer and gastric cancer, and in colorectal cancer (CRC) it facilitates tumor proliferation and invasion. In CRC patients, it is linked with poor prognosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hang Yang et al.
Disease markers, 2015, 640263-640263 (2015-06-13)
Data from The Cancer Genome Atlas (TCGA) show that the ZNF703 gene amplifies and overexpresses in head and neck squamous cell carcinomas (HNSCC). However, the clinical relevance of this observation in HNSCC is unclear. The purpose of this study was
ZNF703 acts as an oncogene that promotes progression in gastric cancer.
Yang G et al
Oncology Reports, 31(4), 1877-1882 (2014)
High expression of ZNF703 independent of amplification indicates worse prognosis in patients with luminal B breast cancer.
Reynisdottir I et al
Cancer Medicine, 2(4), 437-446 (2013)
ZNF703 promotes tumor cell proliferation and invasion and predicts poor prognosis in patients with colorectal cancer.
Ma F et al
Oncology Reports, 32(3), 1071-1077 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service