Skip to Content
Merck
All Photos(6)

Documents

SAB2500422

Sigma-Aldrich

Anti-FOXP3/Scurfin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AIID, Anti-Forkhead box P3, Anti-IPEX, Anti-JM2, Anti-SCURFIN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXP3(50943)

General description

Forkhead box P3 (FOXP3) is a transcription factor and part of the forkhead family. It possesses three α helices and three β strands. The protein has an amino-terminal region, a central zinc finger, a leucine zipper domain and the carboxyl-terminal region has a forkhead domain. The gene encoding this protein is localized on human chromosome Xp11.23.

Immunogen

Peptide with sequence SQRPSRCSNPTPGP from the C Terminus of the protein sequence according to NP_054728.2.

Biochem/physiol Actions

Forkhead box P3 (FOXP3) associates with nuclear factor of activated T cells (NFAT) and nuclear factor κ B (NF-κB). It has a crucial role in the cellular differentiation of regulatory T cells. Alterations in the protein expression have been associated with immunodysregulation, polyendocrinopathy, enteropathy, X-linked (IPEX) syndrome. FOXP3 has a role in tumor growth and cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel pathogenic variants in FOXP3 in fetuses with echogenic bowel and skin desquamation identified by ultrasound.
Louie RJ
American Journal of Medical Genetics. Part A, 173(5) (2017)
The Role of FOXP3 in Human Cancers.
Szylberg L
Anticancer Research (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service