Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500273

Sigma-Aldrich

Anti-CRHR1/CRF-R (AA 250-263) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CRF-R, Anti-CRF1, Anti-CRH-R1h, Anti-Corticotropin releasing hormone receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRHR1(1394)

General description

Corticotropin releasing factor receptor 1 (CRHR1) belongs to the G-protein-coupled receptor superfamily of brain-gut neuropeptides. It is highly expressed in the amygdala, hippocampus and frontal cortex. The gene encoding CRHR1 consists of 13 exons and is localized on human chromosome 17q12-q22.

Immunogen

Peptide with sequence KLYYDNEKCWFGKR from the internal region of the protein sequence according to NP_004373.2.

Biochem/physiol Actions

Corticotropin releasing factor receptor 1 (CRHR1) helps to initiate signal transduction pathways that control various functions like stress, reproduction, immune response and obesity. It blocks the multiplication of endocrine-related cancer cell lines.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The activation of type 1 corticotropin releasing factor receptor (CRF-R1) inhibits proliferation and promotes differentiation of neuroblastoma cells in vitro via p27(Kip1) protein up-regulation and c-Myc mRNA down-regulation.
Pozzoli G
Molecular and Cellular Endocrinology (2015)
Family Economic Hardship, Corticotropin-Releasing Hormone Receptor Polymorphisms, and Depressive Symptoms in Rural African American Youths
Yi-fu Chen
The Journal of Adolescent Health : Official Publication of the Society for Adolescent Medicine (2015)
Association analysis of polymorphisms of the CRHR1 gene with infantile spasms
Guang Yang
Molecular Medicine Reports (2015)
Urocortinergic system in the testes of normal and cryptorchid dogs
Caterina Squillacioti
Annals of Anatomy = Anatomischer Anzeiger : Official Organ of the Anatomische Gesellschaft, 207 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service