Skip to Content
Merck
All Photos(2)

Key Documents

SAB2107342

Sigma-Aldrich

Anti-ITGB1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€416.00

€416.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€416.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€416.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

86 kDa

species reactivity

guinea pig, sheep, human, bovine, goat, rat, dog, horse, mouse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ITGB1(3688)
mouse ... Itgb1(16412)

Immunogen

The immunogen for anti-ITGB1 antibody: synthetic peptide derected towards the C terminal of human ITGB1

Sequence

Synthetic peptide located within the following region: PDIIPIVAGVVAGIVLIGLALLLIWKLLMIIHDRREFAKFEKEKMNAKWD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Zhang et al.
World journal of gastroenterology, 20(23), 7442-7451 (2014-06-27)
To explore the protective effect of bone marrow mesenchymal stem cells (BM MSCs) in the small intestinal mucosal barrier following heterotopic intestinal transplantation (HIT) in a rat model. BM MSCs were isolated from male Lewis rats by density gradient centrifugation
Lauren M Rota et al.
Cancer research, 74(19), 5668-5679 (2014-08-06)
Triple-negative breast cancer (TNBC) is an aggressive disease subtype that, unlike other subtypes, lacks an effective targeted therapy. Inhibitors of the insulin-like growth factor receptor (IGF1R) have been considered for use in treating TNBC. Here, we provide genetic evidence that
J-H Lee et al.
Transplantation proceedings, 46(5), 1633-1637 (2014-06-18)
Liver transplantation is severely limited by donor shortage although it is the only effective treatment for end-stage liver disease. So the best alternative is hepatocyte transplantation. For obtaining human hepatocytes, some stem cells originating from extrahepatic or intraheptic tissues have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service