Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102402

Sigma-Aldrich

Anti-TEAD2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ETF, Anti-TEA domain family member 2, Anti-TEF4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

49 kDa

species reactivity

bovine, human, mouse, horse, rat, rabbit

technique(s)

western blot: suitable

immunogen sequence

GEPRAGAALDDGSGWTGSEEGSEEGTGGSEGAGGDGGPDAEGVWSPDIEQ

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TEAD2(8463)

Immunogen

Peptide region of the protein sequence according to NP_003589.

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Zhou et al.
Oncogene, 36(47), 6518-6530 (2017-08-02)
TEA domain (TEAD) transcription factors are key components of the Hippo-YAP1 signaling pathway, but their functional role and regulatory mechanisms remain unclear. This study aims to comprehensively explore the expression pattern and functional role of TEAD family in gastric carcinogenesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service