Skip to Content
Merck
All Photos(5)

Key Documents

SAB2100474

Sigma-Aldrich

Anti-CPEB4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cytoplasmic polyadenylation element binding protein 4, Anti-KIAA1673

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

80 kDa

species reactivity

human, dog, mouse, guinea pig, rabbit, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPEB4(80315)

General description

Cytoplasmic polyadenylation element-binding protein 4 (CPEB4) is a sequence-specific binding protein that belongs to the CPEB family. It consists of two domains, an unstructured domain, and a RNA binding domain. CPEB4 is expressed in the spinal cord and brain. The CPEB4 gene is mapped on the human chromosome at 5q35.2.

Immunogen

Synthetic peptide directed towards the N terminal region of human CPEB4

Biochem/physiol Actions

Cytoplasmic polyadenylation element-binding protein 4 (CPEB4) plays a key role in translational control during tumor development and modulates various biological processes in tumor progression, including invasion, metastasis, tumor cell growth, and vascularization. It is involved in pathologic angiogenesis, terminal erythroid differentiation, and non-neuronal tissues for cancer progression. Overexpression of the CPEB4 gene is associated with pancreatic ductal adenocarcinoma and glioblastoma.

Sequence

Synthetic peptide located within the following region: DEILGSEKAKSQQQEQQDPLEKQQLSPSPGQEAGILPETEKAKSEENQGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jihae Shin et al.
Scientific reports, 6, 29395-29395 (2016-07-07)
CPEB4 is an RNA binding protein expressed in neuronal tissues including brain and spinal cord. CPEB4 has two domains: one that is structured for RNA binding and one that is unstructured and low complexity that has no known function. Unstructured
Wanming Hu et al.
Medicine, 94(27), e979-e979 (2015-07-15)
CPEB4 plays an important role in cancer progression. However, the clinicopathological significance of CPEB4 expression to glioma and its expression levels in glioma tissues and cell lines are unknown. The present study investigated the potential prognostic value of CPEB4 for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service