Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100073

Sigma-Aldrich

Anti-AGTR1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AG2S, Anti-AT1, Anti-AT1B, Anti-Angiotensin II receptor, type 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€386.00

€386.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€386.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€386.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human, bovine, sheep, guinea pig, rat, rabbit, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGTR1(185)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human AGTR1

Biochem/physiol Actions

Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. It acts through at least two types of receptors. AGTR1 is the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. AGTR1 may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans.Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. It acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. This gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. At least five transcript variants have been described for this gene. Additional variants have been described but their full-length nature has not been determined. The entire coding sequence is contained in the terminal exon and is present in all transcript variants.

Sequence

Synthetic peptide located within the following region: ILNSSTEDGIKRIQDDCPKAGRHNYIFVMIPTLYSIIFVVGIFGNSLVVI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sri Vidya Dangudubiyyam et al.
Biology of reproduction, 107(4), 1072-1083 (2022-07-15)
Epidemiological studies show a strong association between environmental exposure to perfluorooctane sulfonic acid (PFOS) and preeclampsia and fetal growth restriction; however, the underlying mechanisms are unclear. We tested the hypothesis that gestational PFOS exposure leads to pregnancy complications via alterations
Sri Vidya Dangudubiyyam et al.
International journal of molecular sciences, 24(18) (2023-09-28)
Perfluorooctane sulfonic acid (PFOS) exposure during pregnancy induces hypertension with decreased vasodilatory angiotensin type-2 receptor (AT2R) expression and impaired vascular reactivity and fetal weights. We hypothesized that AT2R activation restores the AT1R/AT2R balance and reverses gestational hypertension by improving vascular
Salvatore Panza et al.
Cancers, 13(18) (2021-09-29)
New avenues for glioblastoma therapy are required due to the limited mortality benefit of the current treatments. The renin-angiotensin system (RAS) exhibits local actions and works as a paracrine system in different tissues and tumors, including glioma. The glioblastoma cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service