Skip to Content
Merck
All Photos(4)

Documents

SAB1413285

Sigma-Aldrich

Monoclonal Anti-ANKS4B antibody produced in mouse

new

clone 3E3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ANKS4B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E3, monoclonal

form

buffered aqueous solution

mol wt

73 kDa

species reactivity

human

concentration

~0.3-1 μg/μL (Please refer to lot specific concentration)

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1, kappa

Protein ID accession no.

NM_145865.1

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... ANKS4B(257629)

General description

Mouse monoclonal antibody raised against a full-length recombinant ANKS4B.

Immunogen

ANKS4B (NP_665872.1, 1 a.a. ~ 417 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence

MSTRYHQAASDSYLELLKEATKRDLNLSDEDGMTPTLLAAYHGNLEALEIICSRGGDPDRCDIWGNTPLHFAASNGHAHCVSFLVNFGANIFALDNDLQTPLDAAASREQNECVALLDKAATAQNIMNPKKVTRLKEQAQKNARRQIKECERLQEKHQNKMAHTYSKEESGTLSSSKGTFSRSSPSNASAPGTFGSLSKGIKDTFKIKFKKNKDTAEQVGKEGRSGQRNVMEVFREEEEDSFSGDFKEKLQLSAEEDGSVHHESILNRPGLGSIVFRRNRISSPEDISDSKREFGFKLPSELLQRQGASEADEGAADEEGEENGLKDDLPWDDDEVEWEEDVVDATPLEVFLLSQHLEEFLPIFKREQIDLEALLLCSDEDLQSIQMQLGPRKKVLNAINRRKQVLQQPGQLVDTSL

Physical form

Solution in phosphate buffered saline, pH 7.4

Preparation Note

Purification by Protein A sepharose

Storage and Stability

Storage Instruction: Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maura J Graves et al.
The Journal of biological chemistry, 295(36), 12588-12604 (2020-07-09)
Nutrient-transporting enterocytes interact with their luminal environment using a densely packed collection of apical microvilli known as the brush border. Assembly of the brush border is controlled by the intermicrovillar adhesion complex (IMAC), a protocadherin-based complex found at the tips

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service