Skip to Content
Merck
All Photos(2)

Key Documents

SAB1412264

Sigma-Aldrich

ANTI-SP1 antibody produced in mouse

clone 1G9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

SP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€491.00

€491.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€491.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€491.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G9, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.41 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP1(6667)

General description

Sp1 protein was the first transcription factor to be cloned and characterized. It was first detected in HeLa cells on the basis of its ability to activate the SV40 early promoter transcription. Analysis of structure and function has revealed that Sp1 can be separated into discrete functional domains. The DNA-binding domain consists of three zinc fingers that specifically bind to the GC-box element.

Immunogen

SP1 (NP_612482, 522 a.a. ~ 618 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QLSSMPGLQTINLSALGTSGIQVHPIQGLPLAIANAPGDHGAQLGLHGAGGDGIHDDTAGGEEGENSPDAQPQAGRRTRREACTCPYCKDSEGRGSG*

Biochem/physiol Actions

Sp1 protein was shown to recognize and bind selectively to a GC-rich consensus sequence (GC-box: GGGCGG or CACCC) that presents in the promoter of several important cellular genes, including Simian vacuolating virus 40 (SV40) early, human immunodeficiency virus-1 (HIV-1), platelet-derived growth factor subunit B (PDGF-B), Myc, c-Src etc. In addition to transcription, Sp1 function has been linked to cell growth, cancer and Huntington disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structures of zinc finger domains from transcription factor Sp1. Insights into sequence-specific protein-DNA recognition.
Narayan VA
The Journal of Biological Chemistry, 272(12), 7801-7809 (1997)
Mithramycin is a gene-selective Sp1 inhibitor that identifies a biological intersection between cancer and neurodegeneration.
Sleiman SF
The Journal of Neuroscience, 31(18), 6858-6870 (2011)
Decreased association of the transcription factor Sp1 with genes downregulated in Huntington's disease.
Chen-Plotkin AS
Neurobiology of Disease, 22(2), 233-241 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service