Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411928

Sigma-Aldrich

ANTI-MUS81 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ21012, FLJ44872, MUS81

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€417.00

€417.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€417.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUS81(80198)

General description

MUS81 structure-specific endonuclease subunit is a DNA repair gene, mapped to human chromosome 11q13.1. The encoded protein belongs to the XPF/ ERCC4 (xeroderma pigmentosum, complementation group F) protein family.

Immunogen

MUS81 (NP_079404.2, 1 a.a. ~ 551 a.a) full-length human protein.

Sequence
MAAPVRLGRKRPLPACPNPLFVRWLTEWRDEATRSRHRTRFVFQKALRSLRRYPLPLRSGKEAKILQHFGDGLCRMLDERLQRHRTSGGDHAPDSPSGENSPAPQGRLAEVQDSSMPVPAQPKAGGSGSYWPARHSGARVILLVLYREHLNPNGHHFLTKEELLQRCAQKSPRVAPGSAPPWPALRSLLHRNLVLRTHQPARYSLTPEGLELAQKLAESEGLSLLNVGIGPKEPPGEETAVPGAASAELASEAGVQQQPLELRPGEYRVLLCVDIGETRGGGHRPELLRELQRLHVTHTVRKLHVGDFVWVAQETNPRDPANPGELVLDHIVERKRLDDLCSSIIDGRFREQKFRLKRCGLERRVYLVEEHGSVHNLSLPESTLLQAVTNTQVIDGFFVKRTADIKESAAYLALLTRGLQRLYQGHTLRSRPWGTPGNPESGAMTSPNPLCSLLTFSDFNAGAIKNKAQSVREVFARQLMQVRGVSGEKAAALVDRYSTPASLLAAYDACATPKEQETLLSTIKCGRLQRNLGPALSRTLSQLYCSYGPLT

Biochem/physiol Actions

MUS81 plays a vital role in the metabolism of replication intermediates. In mammals, MUS81 interacts with the non-catalytic subunits essential meiotic structure-specific endonuclease (EME) 1 or EME 2 to form heterodimer complexes. These complexes have the ability to cleave replication forks (RFs) in vitro. Downregulated expression of Mus81 enhances the sensitivity of colon cancer cells to chemotherapeutic drugs by inducing S phase arrest and apoptosis through CHK1 pathway activation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junctions on the road to cancer.
Matthew C Whitby
Nature structural & molecular biology, 11(8), 693-695 (2004-07-29)
M N Boddy et al.
Cell, 107(4), 537-548 (2001-11-24)
Mus81, a fission yeast protein related to the XPF subunit of ERCC1-XPF nucleotide excision repair endonuclease, is essential for meiosis and important for coping with stalled replication forks. These processes require resolution of X-shaped DNA structures known as Holliday junctions.
Heike Duda et al.
Developmental cell, 39(6), 740-755 (2016-12-21)
While DNA replication and mitosis occur in a sequential manner, precisely how cells maintain their temporal separation and order remains elusive. Here, we unveil a double-negative feedback loop between replication intermediates and an M-phase-specific structure-selective endonuclease, MUS81-SLX4, which renders DNA
Fan Wu et al.
Clinics and research in hepatology and gastroenterology, 41(5), 592-601 (2017-03-16)
The inhibition of Mus81, a critical DNA repair gene, is recently related to the chemosensitivity of several human cancer cells such as hepatocellular carcinoma (HCC) cells. However, the role of Mus81 knockdown in chemotherapy response of colon cancer cells remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service