Skip to Content
Merck
All Photos(4)

Key Documents

SAB1411567

Sigma-Aldrich

Anti-FPGT antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

GFPP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€367.00

€367.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€367.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 66.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FPGT(8790)

Related Categories

General description

L-fucose is a key sugar in glycoproteins and other complex carbohydrates since it may be involved in many of the functional roles of these macromolecules, such as in cell-cell recognition. The fucosyl donor for these fucosylated oligosaccharides is GDP-beta-L-fucose. There are two alternate pathways for the biosynthesis of GDP-fucose; the major pathway converts GDP-alpha-D-mannose to GDP-beta-L-fucose. The protein encoded by this gene participates in an alternate pathway that is present in certain mammalian tissues, such as liver and kidney, and appears to function as a salvage pathway to reutilize L-fucose arising from the turnover of glycoproteins and glycolipids. This pathway involves the phosphorylation of L-fucose to form beta-L-fucose-1-phosphate, and then condensation of the beta-L-fucose-1-phosphate with GTP by fucose-1-phosphate guanylyltransferase to form GDP-beta-L-fucose. (provided by RefSeq)

Immunogen

FPGT (NP_003829.2, 1 a.a. ~ 594 a.a) full-length human protein.

Sequence
MAAARDPPEVSLREATQRKLRRFSELRGKLVARGEFWDIVAITAADEKQELAYNQQLSEKLKRKELPLGVQYHVFVDPAGAKIGNGGSTLCALQCLEKLYGDKWNSFTILLIHSGGYSQRLPNASALGKIFTALPLGNPIYQMLELKLAMYIDFPLNMNPGILVTCADDIELYSIGEFEFIRFDKPGFTALAHPSSLTIGTTHGVFVLDPFDDLKHRDLEYRSCHRFLHKPSIEKMYQFNAVCRPGNFCQQDFAGGDIADLKLDSDYVYTDSLFYMDHKSAKMLLAFYEKIGTLSCEIDAYGDFLQALGPGATVEYTRNTSNVIKEESELVEMRQRIFHLLKGTSLNVVVLNNSKFYHIGTTEEYLFYFTSDNSLKSELGLQSITFSIFPDIPECSGKTSCIIQSILDSRCSVAPGSVVEYSRLGPDVSVGENCIISGSYILTKAALPAHSFVCSLSLKMNRCLKYATMAFGVQDNLKKSVKTLSDIKLLQFFGVCFLSCLDVWNLKVTEELFSGNKTCLSLWTARIFPVCSSLSDSVITSLKMLNAVKNKSAFSLNSYKLLSIEEMLIYKDVEDMITYREQIFLEISLKSSLM

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service