Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411331

Sigma-Aldrich

Anti-TBX5 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HOS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 57.7 kDa

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBX5(6910)

General description

T-box 5 (TBX5) is encoded by the gene mapped to human chromosome 12q24.21. The encoded protein belongs to the Brachyury (T) gene family.
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is closely linked to related family member T-box 3 (ulnar mammary syndrome) on human chromosome 12. The encoded protein may play a role in heart development and specification of limb identity. Mutations in this gene have been associated with Holt-Oram syndrome, a developmental disorder affecting the heart and upper limbs. Several transcript variants encoding different isoforms have been described for this gene. (provided by RefSeq)

Immunogen

TBX5 (AAH27942.1, 1 a.a. ~ 518 a.a) full-length human protein.

Sequence
MADADEGFGLAHTPLEPDAKDLPCDSKPESALGAPSKSPSSPQAAFTQQGMEGIKVFLHERELWLKFHEVGTEMIITKAGRRMFPSYKVKVTGLNPKTKYILLMDIVPADDHRYKFADNKWSVTGKAEPAMPGRLYVHPDSPATGAHWMRQLVSFQKLKLTNNHLDPFGHIILNSMHKYQPRLHIVKADENNGFGSKNTAFCTHVFPETAFIAVTSYQNHKITQLKIENNPFAKGFRGSDDMELHRMSRMQSKEYPVVPRSTVRQKVASNHSPFSSESRALSTSSNLGSQYQCENGVSGPSQDLLPPPNPYPLPQEHSQIYHCTKRKEEECSTTDHPYKKPYMETSPSEEDSFYRSSYPQQQGLGASYRTESAQRQACMYASSAPPSEPVPSLEDISCNTWPSMPSYSSCTVTTVQPMDRLPYQHFSAHFTSGPLVPRLAGMANHGSPQLGEGMFQHQTSVAHQPVVRQCGPQTGLQSPGTLQPPEFLYSHGVPRTLSPHQYHSVHGVGMVPEWSDNS

Biochem/physiol Actions

T-box 5 (TBX5) plays a vital role in cardiac differentiation and morphogenesis. It is involved in vertebrate tissue patterning and differentiation. The encoded protein is also implicated in determining limb/wing identity. Mutation in the gene leads to Holt-Oram syndrome, which is associated with the heart and limb malformations.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B G Bruneau et al.
Cell, 106(6), 709-721 (2001-09-27)
Heterozygous Tbx5(del/+) mice were generated to study the mechanisms by which TBX5 haploinsufficiency causes cardiac and forelimb abnormalities seen in Holt-Oram syndrome. Tbx5 deficiency in homozygous mice (Tbx5(del/del)) decreased expression of multiple genes and caused severe hypoplasia of posterior domains
Q Y Li et al.
Nature genetics, 15(1), 21-29 (1997-01-01)
Holt-Oram syndrome is a developmental disorder affecting the heart and upper limb, the gene for which was mapped to chromosome 12 two years ago. We have now identified a gene for this disorder (HOS1). The gene (TBX5) is a member
Chirag Patel et al.
European journal of human genetics : EJHG, 20(8), 863-869 (2012-02-16)
Holt-Oram syndrome (HOS) is a rare autosomal dominant heart-hand syndrome due to mutations in the TBX5 transcription factor. Affected individuals can have structural cardiac defects and/or conduction abnormalities, and exclusively upper limb defects (typically bilateral, asymmetrical radial ray defects). TBX5
J K Takeuchi et al.
Nature, 398(6730), 810-814 (1999-05-11)
Much progress has been made in understanding limb development. Most genes are expressed equally and in the same pattern in the fore- and hindlimbs, which nevertheless develop into distinct structures. The T-box genes Tbx5 and Tbx4, on the other hand
Y Hiroi et al.
Nature genetics, 28(3), 276-280 (2001-06-30)
The cardiac homeobox protein Nkx2-5 is essential in cardiac development, and mutations in Csx (which encodes Nkx2-5) cause various congenital heart diseases. Using the yeast two-hybrid system with Nkx2-5 as the 'bait', we isolated the T-box-containing transcription factor Tbx5; mutations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service