Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410523

Sigma-Aldrich

Anti-GGT1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD224, D22S672, D22S732, GGT, GTG, MGC96892, MGC96904, MGC96963

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGT1(2678)

Related Categories

General description

The enzyme encoded by this gene catalyzes the transfer of the glutamyl moiety of glutathione to a variety of amino acids and dipeptide acceptors. The enzyme is composed of a heavy chain and a light chain, which are derived from a single precursor protein, and is present in tissues involved in absorption and secretion. This enzyme is a member of the gamma-glutamyltransferase protein family, of which many members have not yet been fully characterized and some of which may represent pseudogenes. This gene is classified as type I gamma-glutamyltransferase. Multiple alternatively spliced variants, encoding the same protein, have been identified. (provided by RefSeq)

Immunogen

GGT1 (NP_005256.2, 1 a.a. ~ 569 a.a) full-length human protein.

Sequence
MKKKLVVLGLLAVVLVLVIVGLCLWLPSASKEPDNHVYTRAAVAADAKQCSKIGRDALRDGGSAVDAAIAALLCVGLMNAHSMGIGGGLFLTIYNSTTRKAEVINAREVAPRLAFATMFNSSEQSQKGGLSVAVPGEIRGYELAHQRHGRLPWARLFQPSIQLARQGFPVGKGLAAALENKRTVIEQQPVLCEVFCRDRKVLREGERLTLPQLADTYETLAIEGAQAFYNGSLTAQIVKDIQAAGGIVTAEDLNNYRAELIEHPLNISLGDVVLYMPSAPLSGPVLALILNILKGYNFSRESVESPEQKGLTYHRIVEAFRFAYAKRTLLGDPKFVDVTEVVRNMTSEFFAAQLRAQISDDTTHPISYYKPEFYTPDDGGTAHLSVVAEDGSAVSATSTINLYFGSKVRSPVSGILFNNEMDDFSSPSITNEFGVPPSPANFIQPGKQPLSSMCPTIMVGQDGQVRMVVGAAGGTQITTATALAIIYNLWFGYDVKRAVEEPRLHNQLLPNVTTVERNIDQAVTAALETRHHHTQIASTFIAVVQAIVRTAGGWAAASDSRKGGEPAGY

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service