Skip to Content
Merck
All Photos(1)

Key Documents

SAB1409106

Sigma-Aldrich

Anti-GEM antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIR, MGC26294

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€367.00

€367.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€367.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 32.56 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GEM(2669)

General description

The protein encoded by this gene belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified. (provided by RefSeq)

Immunogen

GEM (ABM84902.1, 1 a.a. ~ 296 a.a) full-length human protein.

Sequence
MTLNNVTMRQGTVGMQPQQQRWSIPADGRHLMVQKEPHQYSHRNRHSATPEDHCRRSWSSDSTDSVISSESGNTYYRVVLIGEQGVGKSTLANIFAGVHDSMDSDCEVLGEDTYERTLMVDGESATIILLDMWENKGENEWLHDHCMQVGDAYLIVYSITDRASFEKASELRIQLRRARQTEDIPIILVGNKSDLVRCREVSVSEGRACAVVFDCKFIETSAAVQHNVKELFEGIVRQVRLRRDSKEKNERRLAYQKRKESMPRKARRFWGKIVAKNNKNMAFKLKSKSCNDLSVL

Biochem/physiol Actions

GEM (GTP binding protein overexpressed in skeletal muscle) is a guanosine triphosphate (GTP)-binding protein. It controls voltage gated Ca2+ channels and cytoskeleton dynamics. Overexpression of GEM causes disruption of stress fiber, changes in cell shape and neurite elongation in interphase cells. In addition, it is also involved in actin remodelling during mitosis. In Timothy syndrome, overexpression of GEM leads to inactivation of RhoA, thereby preventing dendritic retraction. GEM might also be associated with cell invasiveness. In HTLV-1 retrovirus (human T-cell leukemia virus type 1) infection, GEM is involved in cell-to-cell viral transmission.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Andrieu et al.
Carcinogenesis, 35(11), 2503-2511 (2014-09-01)
Gem is a small guanosine triphosphate (GTP)-binding protein within the Ras superfamily, involved in the regulation of voltage-gated calcium channel activity and cytoskeleton reorganization. Gem overexpression leads to stress fiber disruption, actin and cell shape remodeling and neurite elongation in
Sébastien A Chevalier et al.
PLoS pathogens, 10(2), e1003917-e1003917 (2014-03-04)
Efficient HTLV-1 viral transmission occurs through cell-to-cell contacts. The Tax viral transcriptional activator protein facilitates this process. Using a comparative transcriptomic analysis, we recently identified a series of genes up-regulated in HTLV-1 Tax expressing T-lymphocytes. We focused our attention towards

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service