Skip to Content
Merck
All Photos(1)

Key Documents

SAB1409020

Sigma-Aldrich

Monoclonal Anti-FBXO28 antibody produced in mouse

clone 3E5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10766, Fbx28, KIAA0483

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E5, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.74 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBXO28(23219)

General description

Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM

Immunogen

FBXO28 (NP_055991, 191 a.a. ~ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QRAHEVLQELRDISSMAMEYFDEKIVPILKRKLPGSDVSGRLMGSPPVPGPSAALTTMQLFSKQNPSRQEVTKLQQQVKTNGAGVTVLRREISELRTKVQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service