Skip to Content
Merck
All Photos(1)

Documents

SAB1407297

Sigma-Aldrich

Anti-LRRC29 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FBL9, FBXL9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRRC29(26231)

Related Categories

General description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains 9 tandem leucine-rich repeats. Two transcript variants encoding the same protein have been found for this gene. Other variants may occur, but their full-length natures have not been characterized. (provided by RefSeq)

Immunogen

LRRC29 (NP_001004055.1, 1 a.a. ~ 223 a.a) full-length human protein.

Sequence
MYSSGWPAGAAEPRHGRGRELAQALGCMHGAPSQLASLSLAHCSSLKSRPELEHQASGTKDACPEPQGPSLLTLRALQELDLTACSKLTDASLAKVLQFLQLRQLSLSLLPELTDNGLVAVARGCPSLEHLALSHCSRLSDKGWAQAASSWPRLQHLNLSSCSQLIEQTLDAIGQACRQLRVLDVATCPGINMAAVRRFQAQLPQVSCVQSRFVGGADLTLTL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service