Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407070

Sigma-Aldrich

Anti-RUNDC3A antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

RAP2IP, RPIP8

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€367.00

€367.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€367.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RUNDC3A(10900)

General description

The gene RUNDC3A (RUN domain containing 3A) encodes a protein that contains the RUN domain and is mapped to human chromosome 17q21.31. The encoded protein is 400 amino acids long and is mainly expressed in the brain.

Immunogen

RUNDC3A (NP_006686.1, 1 a.a. ~ 405 a.a) full-length human protein.

Sequence
MEASFVQTTMALGLSSKKASSRNVAVERKNLITVCRFSVKTLLEKYTAEPIDDSSEEFVNFAAILEQILSHRFKACAPAGPVSWFSSDGQRGFWDYIRLACSKVPNNCVSSIENMENISTARAKGRAWIRVALMEKRMSEYITTALRDTRTTRRFYDSGAIMLRDEATILTGMLIGLSAIDFSFCLKGEVLDGKTPVVIDYTPYLKFTQSYDYLTDEEERHSAESSTSEDNSPEHPYLPLVTDEDSWYSKWHKMEQKFRIVYAQKGYLEELVRLRESQLKDLEAENRRLQLQLEEAAAQNQREKRELEGVILELQEQLTGLIPSDHAPLAQGSKELTTPLVNQWPSLGTLNGAEGASNSKLYRRHSFMSTEPLSAEASLSSDSQRLGEGTRDEEPWGPIGSSEPN

Biochem/physiol Actions

The gene RUNDC3A (RUN domain containing 3A) encodes a protein that interacts with Rap2, a small GTP-binding protein that belongs to the Ras superfamily, via the RUN domain at the N-terminus. Rap2 may be involved in neuronal functions along with effector molecule, RUNDC3A.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic events associated with progression of pleural malignant mesothelioma.
Ivanov SV
International Journal of Cancer. Journal International Du Cancer, 124, 589-599 (2009)
Identification of a specific effector of the small GTP-binding protein Rap2.
Janoueix-Lerosey I
European Journal of Biochemistry, 252, 290-298 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service