Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406575

Sigma-Aldrich

Anti-USP1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

UBP

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€416.00

€416.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€416.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€416.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~88.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP1(7398)

General description

This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. The protein specifically deubiquitinates a protein in the Fanconi anemia (FA) DNA repair pathway. Alternate transcriptional splice variants have been characterized. (provided by RefSeq)

Immunogen

USP1 (NP_003359.3, 1 a.a. ~ 785 a.a) full-length human protein.

Sequence
MPGVIPSESNGLSRGSPSKKNRLSLKFFQKKETKRALDFTDSQENEEKASEYRASEIDQVVPAAQSSPINCEKRENLLPFVGLNNLGNTCYLNSILQVLYFCPGFKSGVKHLFNIISRKKEALKDEANQKDKGNCKEDSLASYELICSLQSLIISVEQLQASFLLNPEKYTDELATQPRRLLNTLRELNPMYEGYLQHDAQEVLQCILGNIQETCQLLKKEEVKNVAELPTKVEEIPHPKEEMNGINSIEMDSMRHSEDFKEKLPKGNGKRKSDTEFGNMKKKVKLSKEHQSLEENQRQTRSKRKATSDTLESPPKIIPKYISENESPRPSQKKSRVKINWLKSATKQPSILSKFCSLGKITTNQGVKGQSKENECDPEEDLGKCESDNTTNGCGLESPGNTVTPVNVNEVKPINKGEEQIGFELVEKLFQGQLVLRTRCLECESLTERREDFQDISVPVQEDELSKVEESSEISPEPKTEMKTLRWAISQFASVERIVGEDKYFCENCHHYTEAERSLLFDKMPEVITIHLKCFAASGLEFDCYGGGLSKINTPLLTPLKLSLEEWSTKPTNDSYGLFAVVMHSGITISSGHYTASVKVTDLNSLELDKGNFVVDQMCEIGKPEPLNEEEARGVVENYNDEEVSIRVGGNTQPSKVLNKKNVEAIGLLGGQKSKADYELYNKASNPDKVASTAFAENRNSETSDTTGTHESDRNKESSDQTGINISGFENKISYVVQSLKEYEGKWLLFDDSEVKVTEEKDFLNSLSPSTSPTSTPYLLFYKKL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiguo Niu et al.
Journal of Cancer, 11(23), 6992-7000 (2020-10-31)
Breast cancer is one of the most common malignancies worldwide, while the luminal types (ERα positive) accounts for two third of all breast cancer cases. Although ERα positive breast cancer could be effective controlled by endocrine therapy, most of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service