Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406286

Sigma-Aldrich

Anti-PPP2R1B antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC26454, PR65B

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€367.00

€367.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€367.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~66.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP2R1B(5519)

General description

This gene encodes a constant regulatory subunit of protein phosphatase 2. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. This gene encodes a beta isoform of the constant regulatory subunit A. Defects in this gene could be the cause of some lung and colon cancers. At least two transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

PPP2R1B (NP_002707, 1 a.a. ~ 601 a.a) full-length human protein.

Sequence
MAGASELGTGPGAAGGDGDDSLYPIAVLIDELRNEDVQLRLNSIKKLSTIALALGVERTRSELLPFLTDTIYDEDEVLLALAEQLGNFTGLVGGPDFAHCLLPPLENLATVEETVVRDKAVESLRQISQEHTPVALEAYFVPLVKRLASGDWFTSRTSACGLFSVCYPRASNAVKAEIRQQFRSLCSDDTPMVRRAAASKLGEFAKVLELDSVKSEIVPLFTSLASDEQDSVRLLAVEACVSIAQLLSQDDLETLVMPTLRQAAEDKSWRVRYMVADRFSELQKAMGPKITLNDLIPAFQNLLKDCEAEVRAAAAHKVKELGENLPIEDRETIIMNQILPYIKELVSDTNQHVKSALASVIMGLSTILGKENTIEHLLPLFLAQLKDECPDVRLNIISNLDCVNEVIGIRQLSQSLLPAIVELAEDAKWRVRLAIIEYMPLLAGQLGVEFFDEKLNSLCMAWLVDHVYAIREAATNNLMKLVQKFGTEWAQNTIVPKVLVMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVLALA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service