Skip to Content
Merck
All Photos(1)

Key Documents

SAB1405455

Sigma-Aldrich

Anti-AMPH antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AMPH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~76.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AMPH(273)

General description

This gene encodes a protein associated with the cytoplasmic surface of synaptic vesicles. A subset of patients with stiff-man syndrome who were also affected by breast cancer are positive for autoantibodies against this protein. Alternate splicing of this gene results in two transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences have not been determined. (provided by RefSeq)

Immunogen

AMPH (AAH34376.1, 1 a.a. ~ 695 a.a) full-length human protein.

Sequence
MADIKTGIFAKNVQKRLNRAQEKVLQKLGKADETKDEQFEEYVQNFKRQEAEGTRLQRELRGYLAAIKGMQEASMKLTESLHEVYEPDWYGREDVKMVGEKCDVLWEDFHQKLVDGSLLTLDTYLGQFPDIKNRIAKRSRKLVDYDSARHHLEALQSSKRKDESRISKAEEEFQKAQKVFEEFNVDLQEELPSLWSRRVGFYVNTFKNVSSLEAKFHKEIAVLCHKLYEVMTKLGDQHADKAFTIQGAPSDSGPLRIAKTPSPPEEPSPLPSPTASPNHTLAPASPAPARPRSPSQTRKGPPVPPLPKVTPTKELQQENIISFFEDNFVPEISVTTPSQNEVPEVKKEETLLDLDFDPFKPEVTPAGSAGVTHSPMSQTLPWDLWTTSTDLVQPASGGSFNGFTQPQDTSLFTMQTDQSMICNLAESEQAPPTEPKAEEPLAAVTPAVGLDLGMDTRAEEPVEEAVIIPGADADAAVGTLVSAAEGAPGEEAEAETATVPAGEGVSLEEAKIGTETTEGAESAQPEAEELEATVPQEKVIPSVVIEPASNHEEEGENEITIGAEPKETTEDAAPPGPTSETPELATEQKPIQDPQPTPSAPAMGAADQLASAREASQELPPGFLYKVETLHDFEAANSDELTLQRGDVVLVVPSDSEADQDAGWLVGVKESDWLQYRDLATYKGLFPENFTRRLD

Application

Anti-AMPH antibody produced in mouse is suitable for western blot.

Biochem/physiol Actions

AMPH (Amphiphysin) is involved in several pleotropic functions including endocytosis, actin function, and cell growth control. It plays an important role in synaptic vesicle endocytosis of axon terminals. Alteration in AMPH gene is associated with heritable disorders of the nervous system, endocrine tissues and male fertility. It also has a correlation with a rare neurologic autoimmune disease, paraneoplastic Stiff-Man syndrome. It is also involved in neoplastic growth cone dynamics.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Floyd et al.
Molecular medicine (Cambridge, Mass.), 4(1), 29-39 (1998-03-26)
Amphiphysin I is a 128 kD protein highly concentrated in nerve terminals, where it has a putative role in endocytosis. It is a dominant autoantigen in patients with stiff-man syndrome associated with breast cancer, as well as in other paraneoplastic
R Yamamoto et al.
Human molecular genetics, 4(2), 265-268 (1995-02-01)
Amphiphysin is a protein peripherally associated with synaptic vesicles. It is expressed in many neurons, certain endocrine cell types, and spermatocytes. Autoantibodies against amphiphysin occur in patients afflicted with a rare neurologic autoimmune disease, paraneoplastic Stiff-Man syndrome. To provide a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service