Skip to Content
Merck
All Photos(7)

Key Documents

SAB1403611

Sigma-Aldrich

Monoclonal Anti-BMP7 antibody produced in mouse

clone S52, purified immunoglobulin, buffered aqueous solution

Synonym(s):

OP-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1627


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1627

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S52, monoclonal

form

buffered aqueous solution

mol wt

antigen ~73.52 kDa

species reactivity

mouse, human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP7(655)

General description

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development and possible bone inductive activity. (provided by RefSeq)

Immunogen

BMP7 (AAH08584, 1 a.a. ~ 431 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MHVRSLRAAAPHSFVALWAPLFLLRSALADFSLDNEVHSSFIHRRLRSQERREMQREILSILGLPHRPRPHLQGKHNSAPMFMLDLYNAMAVEEGGGPGGQGFSYPYKAVFSTQGPPLASLQDSHFLTDADMVMSFVNLVEHDKEFFHPRYHHREFRFDLSKIPEGEAVTAAEFRIYKDYIRERFDNETFRISVYQVLQEHLGRESDLFLLDSRTLWASEEGWLVFDITATSNHWVVNPRHNLGLQLSVETLDGQSINPKLAGLIGRHGPQNKQPFMVAFFKATEVHFRSIRSTGSKQRSQNRSKTPKNQEALRMANVAENSSSDQRQACKKHELYVSFRDLGWQDWIIAPEGYAAYYCEGECAFPLNSYMNATNHAIVQTLVHFINPETVPKPCCAPTQLNAISVLYFDDSSNVILKKYRNMVVRACGCH

Biochem/physiol Actions

As implied by their name, bone morphogenetic proteins (BMPs) promote and regulate bone development and growth. BMP-7 induces apoptosis in B-cells. It has a role in organ development and may have a role in liver regeneration. Mutations in the gene encoding the protein have been shown to affect the development of teeth, palate and other orofacial complexes.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caixia Yuan et al.
Experimental and therapeutic medicine, 17(4), 2547-2556 (2019-03-25)
Bone morphogenetic protein (BMP) expression has been observed in the uterus in previous studies. However, the influence of BMP7 on blastocyst implantation remains unclear. Blastocysts first act on luminal endometrial epithelial cells during implantation. The purpose of the present study
Applications of small molecule BMP inhibitors in physiology and disease.
Hong CC and Yu PB
Cytokine & Growth Factor Reviews, 20(5-6), 409-418 (2009)
BMP7 expression correlates with secondary drug resistance in mantle cell lymphoma.
Camara-Clayette V
PLoS ONE, 8(9), e73993-e73993 (2013)
BMP7 Gene involved in nonsyndromic orofacial clefts in Western Han Chinese.
Yu Q
Medicina Oral, patologia Oral y cirugia Bucal, 20(3), e298-e304 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service