Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403506

Sigma-Aldrich

Monoclonal Anti-MAP3K15 antibody produced in mouse

clone 1H7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ16518, bA723P2.3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.67 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Mitogen-activated protein kinase kinase kinase 15 (MAP3K15) belongs to the group of apoptosis signal-regulating kinases. It is expressed in the kidney and the gene encoding it is localized on human chromosome Xp22.12.

Immunogen

MAP3K15 (NP_001001671, 691 a.a. ~ 786 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LYHLQLKLKSNCITENPAGPYGQRTDKELIDWLRLQGADAKTIEKIVEEGYTLSDILNEITKEDLRYLRLRGGLLCRLWSAVSQYRRAQEASETKD

Biochem/physiol Actions

Mitogen-activated protein kinase kinase kinase 15 (MAP3K15) modulates blood pressure in the kidney. It has a role in signal transduction cascades related to apoptotic cell death initiated due to cellular stress. ASK3 has been shown to protect HeLa cells against cytotoxicity induced by oxidative stress, antibodies or proteins.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ASK3, a novel member of the apoptosis signal-regulating kinase family, is essential for stress-induced cell death in HeLa cells.
Kaji T
Biochemical and Biophysical Research Communications (2010)
ASK3 responds to osmotic stress and regulates blood pressure by suppressing WNK1-SPAK/OSR1 signaling in the kidney.
Naguro I
Nature Communications (2012)
MAP7D2 is a brain expressing X-linked maternal imprinted gene in humans
Yo NIIDA
Natural Products Journal (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service