Skip to Content
Merck
All Photos(1)

Key Documents

SAB1402677

Sigma-Aldrich

Monoclonal Anti-ABCA1 antibody produced in mouse

clone 1H4, ascites fluid

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€416.00

€416.00


Please contact Customer Service for Availability


Select a Size

Change View
200 μL
€416.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€416.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H4, monoclonal

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCA1(19)

General description

ATP binding cassette subfamily A member 1 (ABCA1) is encoded by the gene mapped to human chromosome 9q31.1. The encoded protein belongs to the ATP-binding cassette (ABC)1 family of membrane transporters.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. With cholesterol as its substrate, this protein functions as a cholesteral efflux pump in the cellular lipid removal pathway. Mutations in this gene have been associated with Tangier′s disease and familial high-density lipoprotein deficiency. (provided by RefSeq)

Immunogen

ABCA1 (NP_005493, 86 a.a. ~ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TPGEAPGVVGNFNKSIVARLFSDARRLLLYSQKDTSMKDMRKVLRTLQQIKKSSSNLKLQDFLVDNETFSGFLYHNLSLPKSTVDKMLRADVILHKVFLQ

Application

Monoclonal Anti-ABCA1 antibody produced in mouse has been used in confocal laser scanning microscopy and Western blot technique.

Biochem/physiol Actions

ATP binding cassette subfamily A member 1 (ABCA1) functions as a phospholipid and/or cholesterol transporter. ABCA1 interacts with its ligand apolipoprotein to regulate cholesterol trafficking. Mutation in the gene is associated with the development of Tangier disease and familial hypoalipoproteinemia.

Physical form

Clear solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Yan Liu et al.
Journal of biochemistry, 155(1), 35-42 (2014-01-08)
ATP-binding cassette transporter A1 (ABCA1) and CD36, type B scavenger receptor, function as the key mediators of macrophages cholesterol efflux and intake, respectively. However, their contribution to development of foam cells still remains uncertain. We here examined the effects of
Identification and functional analysis of a naturally occurring E89K mutation in the ABCA1 gene of the WHAM chicken.
Attie A D, et al.
Journal of Lipid Research, 43(10), 1610-1617 (2002)
ABCA1 Is the cAMP-inducible Apolipoprotein Receptor That Mediates Cholesterol Secretion from Macrophages
Oram J F, et al.
The Journal of Biological Chemistry, 275(44), 34508-34511 (2000)
Imbalanced response of ATP-binding cassette transporter A1 and CD36 expression to increased oxidized low-density lipoprotein loading contributes to the development of THP-1 derived foam cells
Liu H Y, et al.
Journal of Biochemistry, 155(1), 35-42 (2014)
Transcriptional Profiling Identifies Two Members of the ATP-binding Cassette Transporter Superfamily Required for Sterol Uptake in Yeast
Wilcox L J, et al.
The Journal of Biological Chemistry, 277(36), 32466-32472 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service