Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402530

Sigma-Aldrich

Monoclonal Anti-POLQ antibody produced in mouse

clone 1C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp781A0112, POLH, PRO0327

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.1 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POLQ(10721)

Immunogen

POLQ (NP_006587, 2572 a.a. ~ 2680 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DGFVQTILGRRRYLPGIKDNNPYRKAHAERQAINTIVQGSAADIVKIATVNIQKQLETFHSTFKSHGHREGMLQSDRTGLSRKRKLQGMFCPIRGGFFILQLHDELLYE

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinrui Rao et al.
Cancer science, 114(5), 1943-1957 (2023-01-16)
Radioresistance remains a major obstacle to efficacious radiotherapy in non-small-cell lung cancer (NSCLC). DNA replication proteins are novel targets for radiosensitizers. POLQ is a DNA polymerase involved in DNA damage response and repair. We found that POLQ is overexpressed in
Yichen He et al.
Journal of cancer research and clinical oncology, 150(2), 30-30 (2024-01-25)
As the first identified m6A demethylase, FTO has been implicated in the progression of various cancers. However, the specific mechanism of FTO in clear cell renal cell carcinoma (ccRCC) remains incompletely understood. In this study, we aimed to explore the
Daniel J Laverty et al.
Nature communications, 15(1), 5294-5294 (2024-06-22)
Determining the balance between DNA double strand break repair (DSBR) pathways is essential for understanding treatment response in cancer. We report a method for simultaneously measuring non-homologous end joining (NHEJ), homologous recombination (HR), and microhomology-mediated end joining (MMEJ). Using this
Jia Zhou et al.
Nature cancer, 2(6), 598-610 (2021-06-29)
DNA polymerase theta (POLθ) is synthetic lethal with Homologous Recombination (HR) deficiency and thus a candidate target for HR-deficient cancers. Through high-throughput small molecule screens we identified the antibiotic Novobiocin (NVB) as a specific POLθ inhibitor that selectively kills HR-deficient
Ruben Schep et al.
Molecular cell, 81(10), 2216-2230 (2021-04-14)
DNA double-strand break (DSB) repair is mediated by multiple pathways. It is thought that the local chromatin context affects the pathway choice, but the underlying principles are poorly understood. Using a multiplexed reporter assay in combination with Cas9 cutting, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service