Skip to Content
Merck
All Photos(2)

Documents

SAB1402512

Sigma-Aldrich

Monoclonal Anti-PCGF3, (C-terminal) antibody produced in mouse

clone 1F1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686D20235, DONG1, FLJ36550, FLJ43813, MGC129615, MGC40413, RNF3, RNF3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCGF3(10336)

Related Categories

General description

The protein encoded by this gene contains a C3HC4 type RING finger, which is a motif known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. (provided by RefSeq)

Immunogen

PCGF3 (NP_006306, 133 a.a. ~ 242 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SNKEAAEEKPEEDNDYHRSDEQVSICLECNSSKLRGLKRKWIRCSAQATVLHLKKFIAKKLNLSSFNELDILCNEEILGKDHTLKFVVVTRWRFKKAPLLLHYRPKMDLL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service