Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402228

Sigma-Aldrich

Monoclonal Anti-HOXA6 antibody produced in mouse

clone 3A6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HOX1, HOX1.2, HOX1B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXA6(3203)

General description

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. (provided by RefSeq)

Immunogen

HOXA6 (NP_076919, 31 a.a. ~ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GYDALRPFPASYGASSLPDKTYTSPCFYQQSNSVLACNRASYEYGASCFYSDKDLSGASPSGSGKQRGPGDYLHFSPEQQYKPDSSSGQGKALHDEGADR

Biochem/physiol Actions

The members of HOX (Homeobox) gene family clusters with each other to form a complex. In HOXA6 (homeobox A6), there are eight homeoboxes of 90kb DNA in the cluster. It is expressed in the human adult small intestinal mucosa. It acts as transcription factors in defining gradients of cellular differentiation. It mainly functions in hemopoietic progenitor cell development, embryonic and fetal development.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Acampora et al.
Nucleic acids research, 17(24), 10385-10402 (1989-12-25)
We report the identification of 10 new human homeobox sequences. Altogether, we have isolated and sequenced 30 human homeoboxes clustered in 4 chromosomal regions called HOX loci. HOX1 includes 8 homeoboxes in 90 kb of DNA on chromosome 7. HOX2
E Boncinelli et al.
Genome, 31(2), 745-756 (1989-01-01)
We report the genomic organization of 20 human class I homeoboxes and the predicted primary sequence of the encoded homeodomains. These homeoboxes are clustered in four complex HOX loci on chromosomes 2, 7, 12, and 17. The homeoboxes of one
J R Walters et al.
Gastroenterology, 113(2), 472-477 (1997-08-01)
Different digestive enzymes and transporters are present in the duodenum, jejunum, and ileum, but the factors determining region-specific gene expression are not yet understood. Homeobox transcription factors are important in defining gradients of cellular differentiation. The aim of this study
D Duboule et al.
The EMBO journal, 8(5), 1497-1505 (1989-05-01)
This paper reports the cloning of the fourth major murine homeogene complex, HOX-5. The partial characterization of this gene cluster revealed the presence of two novel genes (Hox-5.2, Hox-5.3) located at the 5' extremity of this complex. In situ hybridization
Glenda J Dickson et al.
Experimental hematology, 37(3), 322-333 (2009-01-23)
Hemopoietic progenitor cells express clustered homeobox (Hox) genes in a pattern characteristic of their lineage and stage of differentiation. In general, HOX expression tends to be higher in more primitive and lower in lineage-committed cells. These trends have led to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service