Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401833

Sigma-Aldrich

Monoclonal Anti-NUP85 antibody produced in mouse

clone 3E9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ12549, Nup75

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€416.00

€416.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€416.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€416.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUP85(79902)

General description

Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160. (provided by RefSeq)

Immunogen

NUP85 (AAH00697, 1 a.a. ~ 656 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEELDGEPTVTLIPGVNSKKNQMYFDWGPGEMLVCETSFNKKEKSEMVPSCPFIYIIRKDVDVYSQILRKLFNESHGIFLGLQRIDEELTGKSRKSQLVRVSKNYRSVIRACMEEMHQVAIAAKDPANGRQFSSQVSILSAMELIWNLCEILFIEVAPAGPLLLHLLDWVRLHVCEVDSLSADVLGSENPSKHDSFWNLVTILVLQGRLDEARQMLSKEADASPASAGICRIMGDLMRTMPILSPGNTQTLTELELKWQHWHEECERYLQDSTFATSPHLESLLKIMLGDEAALLEQKELLSNWYHFLVTRLLYSNPTVKPIDLHYYAQSSLDLFLGGESSPEPLDNILLAAFEFDIHQVIKECSIALSNWWFVAHLTDLLDHCKLLQSHNLYFGSNMREFLLLEYASGLFAHPSLWQLGVDYFDYCPELGRVSLELHIERIPLNTEQKALKVLRICEQRQMTEQVRSICKILAMKAVRNNRLGSALSWSIRAKDAAFATLVSDRFLRDYCERGCFSDLDLIDNLGPAMMLSDRLTFLGKYREFHRMYGEKRFADAASLLLSLMTSRIAPRSFWMTLLTDALPLLEQKQVIFSAEQTYELMRCLEDLTSRRPVHGESDTEQLQDDDIETTKVEMLRLSLARNLARAIIREGSLEGS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service