Skip to Content
Merck
All Photos(2)

Documents

SAB1401759

Sigma-Aldrich

Monoclonal Anti-KIF16B antibody produced in mouse

clone 2B5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

C20orf23, KISC20ORF, SNX23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIF16B(55614)

Immunogen

KIF16B (AAH34984, 65 a.a. ~ 174 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DDLKDPIKISIPRYVLCGQGKDAHFEFEVKLAALEFPPKKLFGNKDERVIAERRSHLEKYLRDFFSVMLQSATSPLHINKVGLTLSKHTICEFSPFFKKGVFDYSSHGTG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Beaudet et al.
The Journal of biological chemistry, 300(6), 107323-107323 (2024-04-28)
Organelles and vesicular cargoes are transported by teams of kinesin and dynein motors along microtubules. We isolated endocytic organelles from cells at different stages of maturation and reconstituted their motility along microtubules in vitro. We asked how the sets of motors
Emily N P Prowse et al.
Biophysical journal, 122(7), 1168-1184 (2023-02-12)
Huntingtin (HTT) is a scaffolding protein that recruits motor proteins to vesicular cargoes, enabling it to regulate kinesin-1, dynein, and myosin-VI-dependent transport. To maintain the native stoichiometry of HTT with its interacting partners, we used CRISPR/Cas9 to induce a phosphomimetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service