Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401292

Sigma-Aldrich

Monoclonal Anti-POLR2C antibody produced in mouse

clone 1C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

RPB3, RPB31, hRPB33, hsRPB3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POLR2C(5432)

General description

This gene encodes the third largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains a cysteine rich region and exists as a heterodimer with another polymerase subunit, POLR2J. These two subunits form a core subassembly unit of the polymerase. A pseudogene has been identified on chromosome 21. (provided by RefSeq)

Immunogen

POLR2C (AAH28157, 1 a.a. ~ 275 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPYANQPTVRITELTDENVKFIIENTDLAVANSIRRVFIAEVPIIAIDWVQIDANSSVLHDEFIAHRLGLIPLISDDIVDKLQYSRDCTCEEFCPECSVEFTLDVRCNEDQTRHVTSRDLISNSPRVIPVTSRNRDNDPNDYVEQDDILIVKLRKGQELRLRAYAKKGFGKEHAKWNPTAGVAFEYDPDNALRHTVYPKPEEWPKSEYSELDEDESQAPYDPNGKPERFYYNVESCGSLRPETIVLSALSGLKKKLSDLQTQLSHEIQSDVLTIN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service