Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306751

Sigma-Aldrich

ANTI-TRIM6 (N-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

RING finger protein 89, RNF89, TRIM6, Tripartite motif-containing protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€388.00

€388.00


Please contact Customer Service for Availability


Select a Size

Change View
400 μL
€388.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€388.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56400 Da

species reactivity

human

technique(s)

western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM6(117854)

Related Categories

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifeng Zhang et al.
Turkish journal of medical sciences, 53(5), 1032-1044 (2023-02-28)
Hepatocellular carcinoma (HCC) ranks among the most prevalent malignancies worldwide and the third leading cause of cancer-related death. The TRIM (tripartite motif-containing) protein family members had been reported to be involved in carcinogenesis and tumor progression. Here we aimed to
Guangwei Zeng et al.
Aging, 11(11), 3536-3550 (2019-06-07)
Cardiomyocyte apoptosis is a major cause of myocardial ischemia/reperfusion (MI/R) injury, in which the activation of the signal transducer and activator of transcription 1 (STAT1) plays an important role. The E3-ubiquitin ligase TRIM6 has been implicated in regulating STAT1 activity
Adam Hage et al.
Cell reports, 38(10), 110434-110434 (2022-03-10)
Type I interferons (IFN-I) are essential to establish antiviral innate immunity. Unanchored (or free) polyubiquitin (poly-Ub) has been shown to regulate IFN-I responses. However, few unanchored poly-Ub interactors are known. To identify factors regulated by unanchored poly-Ub in a physiological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service