Skip to Content
Merck
All Photos(2)

Key Documents

SAB1301592

Sigma-Aldrich

ANTI-DIO3 (C-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

5DIII, DIO3, DIOIII, ITDI3, TXDI3, Type 3 DI, Type III iodothyronine deiodinase, Type-III 5′-deiodinase

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€457.00

€457.00


Please contact Customer Service for Availability


Select a Size

Change View
400 μL
€457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€457.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31451 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:250-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DIO3(1735)

General description

Iodothyronine deiodinase 3 (DIO3) is highly expressed in central nervous system, fetal liver and placenta. DIO3 is a selenoenzyme with insertion sequence element containing selenocysteine in the 3′UTR (untranslated region). In human chromosome, the gene DIO3 is localized on 14q32.31.

Biochem/physiol Actions

Iodothyronine deiodinase 3 (DIO3) regulates the serum level of thyroid hormone. DIO3 removes iodine from the inner ring of T4 (thyroxine) and T3 (triiodothyronine) and converts to its inactive forms. The major role of DIO3 is to protect exposure of fetal tissues to high level of T3 throughout gestation. Elevated expression of DIO3 is associated with infantile hepatic hemangiomas (IHH). Alterations in the thyroid hormone exposure during developmental stages may cause phenotypes similar to uniparental disomy of chromosome 14. The lack of DIO3 leads to neonatal thyrotoxicosis, followed by hypothyroidism. DIO3 is crucial for auditory development. Mutations in DIO3 leads to deafness due to immature cochlear differentiation. DIO3 protects the immature cones by regulating the hormonal exposure, facilitating cone survival and opsins patterning.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The genetics and functional analysis of primary osteoarthritis susceptibility
Reynard LN and Loughlin J
Expert Reviews in Molecular Medicine, 15(2), e2-e2 (2013)
Type 3 deiodinase, a thyroid-hormone-inactivating enzyme, controls survival and maturation of cone photoreceptors
Ng L, et al.
The Journal of Neuroscience, 30(9), 3347-3357 (2010)
Increased type 3 iodothyronine deiodinase activity in a regrown hepatic hemangioma with consumptive hypothyroidism
Bessho K, et al.
European Journal of Pediatrics, 169(2), 215-215 (2010)
A protective role for type 3 deiodinase, a thyroid hormone-inactivating enzyme, in cochlear development and auditory function
Ng L, et al.
Endocrinology, 150(4), 1952-1960 (2008)
Genomic imprinting of DIO3, a candidate gene for the syndrome associated with human uniparental disomy of chromosome 14
Martinez ME, et al.
European Journal of Human Genetics, 24(11), 1617-1617 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service