Skip to Content
Merck
All Photos(1)

Documents

SAB1300668

Sigma-Aldrich

Anti-SULT2A (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Alcohol sulfotransferase, Anti-DHEA-ST, Anti-HST, Anti-Hydroxysteroid Sulfotransferase, Dehydroepiandrosterone sulfotransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULT2A(6822)

General description

One of the major roles of the sulfotransferases (ST) in the metabolism of drugs and endogenous compounds is the conversion of these substances into more hydrophilic water-soluble sulfate conjugates that can be easily excreted. Sulfation may also play a regulatory role for many endogenous compounds, such as steroids and neurotransmitters, by altering the biologic properties of these compounds. Otterness et al. (1992), Kong et al. (1992), and Comer et al. (1993) reported the cloning of cDNAs encoding liver dehydroepiandrosterone (DHEA) sulfotransferase. The predicted protein has 285 amino acids. Although Northern blot analysis of human liver RNA detected transcripts of 3 different sizes, Southern blot analysis of human DNA suggested that only 1 gene is present in the genome. This gene has an important role in the sulfation of both bile acids and steroids in the liver and adrenals. The human adrenal form of this enzyme is physically, immunologically, and kinetically similar, perhaps identical, to the liver form. Dehydroepiandrosterone sulfate is quantitatively one of the major steroids secreted from the adrenal cortex.

Immunogen

SULT2A (NP_003158, 9-46)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human SULT2A.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service