Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300557

Sigma-Aldrich

Anti-GJA3 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Connexin-46, Anti-Cx46, Anti-Gap junction α-3 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€318.00

€318.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€318.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€318.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GJA3(2700)

General description

GJA3, an integral membrane protein, belong to the connexin family, alpha-type (group II) subfamily. One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell. A connexon is composed of a hexamer of connexins. This particular connexin is a component of lens fiber gap junctions, can form both junctional and nonjunctional (′hemi-′) channels. Defects in GJA3 are the cause of zonular pulverulent cataract type 3 (CZP3), a form of autosomal dominant congenital cataract.

Immunogen

GJA3 (NP_068773, 112-148)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-term region of human GJA3.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gergo Kiszner et al.
Cancers, 11(2) (2019-02-06)
The incidence of malignant melanoma, one of the deadliest cancers, continues to increase. Here we tested connexin (Cx) expression in primary melanocytes, melanoma cell lines and in a common nevus, dysplastic nevus, and thin, thick, and metastatic melanoma tumor progression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service